Search
| Número de catálogo | Cantidad |
|---|---|
| R24229 | 40 tests/kit |
Thermo Scientific™ Remel Mycoplasma Pneumoniae IgG/IgM Antibody Test is an enzyme-linked immunobinding assay used for the qualitative simultaneous detection of IgG and IgM antibodies to Mycoplasma pneumoniae in human serum or plasma.