Search Thermo Fisher Scientific
Remel ImmunoWELL EBNA IgG is an EIA microwell test used for qualitative detection of IgG antibodies to Epstein-Barr Virus nuclear antigen-1 in serum.
Packaging Type | 1 Kit |
Quantity | 96 Wells |
Description | ImmunoWELL™ EBNA IgG |
Detectable Analytes | IgG antibodies to Epstein-Barr Virus nuclear antigen-1 in serum |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R243230 | Full specifications | Each | 96 Wells | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|