This product is only available through your local Thermo Scientific supplier. Learn where to buy >
Digestion Solution for MagJET gDNA Kit
Catalog number: R2452
Thermo Scientific™  Related applications: DNA Extraction
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific MagJet Digestion Solution is a component of several MagJET nucleic acid purification kits and may be purchased separately. Please consult kit protocols to confirm catalog numbers for re-ordering the appropriate kit-specific components.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer Type: Digestion Buffer
Product Line: MagJET
Product Size: 90 mL
Documents
Manuals & protocols