ProSpecT™ Giardia Microplate Assay
Thermo Scientific™
ProSpecT™ Giardia Microplate Assay
Rapidly detect Giardia specific antigen from human fecal samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R245802424 Tests/Kit
R245809696 Tests/Kit
Catalog number R2458024
Price (USD)
-
Quantity:
24 Tests/Kit
Rapidly detect Giardia specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Giardia Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Giardia. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Giardia is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Giardia provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Specifications
DescriptionProSpecT Giardia Microplate Assay
For Use With (Application)Enzyme Immunoassay Detection
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity24 Tests/Kit
Sample TypeFecal
TypeGiardia Microplate Assay
Detectable AnalytesGiardia Cyst Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C