Flu A and B Control Swab
Thermo Scientific™
Flu A and B Control Swab
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.
Have Questions?
Número de catálogoCantidad
R24600340 hisopos
Número de catálogo R246003
Precio (MXN)
-
Cantidad:
40 hisopos
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.
These kits contain positive and negative controls to monitor test performance.
Human influenza A and B viruses cause seasonal epidemics of disease (known as the flu season) almost every winter. Flu A is known to cause flu pandemics. The Influenza A and B test is performed for the qualitative detection and differentiation of Influenza A virus, including H5N1 and H1N1, and Influenza B virus in nasal/throat/nasopharyngeal swab or nasopharyngeal aspirate.
This kit contains Flu A control dry swab containing inactivated influenza A antigen and Flu B control dry swab containing inactivated influenza B antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Especificaciones
DescripciónXpect Flu A and B Control Swab
Incluye20 Flu A+/Flu B- control swabs and 20 Flu A-/Flu B+ control swabs
Cantidad40 hisopos
TipoControl
Analitos detectablesInfluenza
Unit SizeEach
Contenido y almacenamiento
De 2 °C a 8 °C