Remel™ Xpect™ RSV Control Kit
Remel™ Xpect™ RSV Control Kit
Remel™ Xpect™ RSV Control Kit
Thermo Scientific™
Remel™ Xpect™ RSV Control Kit
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
Have Questions?
Catalog NumberQuantity
R2460122 Vials
Catalog number R246012
Price (USD)
-
Quantity:
2 Vials
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
  • Method: Chromatographic immunoassay
  • Samples collected from nasopharyngeal specimens in neonatal and paediatric patients
Ordering Alerts:
Six positive and negative tests can be run from 1 set of controls.
Includes:
1 vial positive control and 1 vial negative control. 1mL/Vial.
Specifications
Description6 Tests
Includes1 Vial positive control and 1 Vial negative control. 1mL/Vial.
Quantity2 Vials
TypeControl
Unit SizeEach