Remel ImmunoDOT Mono IgG is an EIA membrane test that simultaneously detects IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum.
Description | ImmunoDOT™ Mono IgG |
Detectable Analytes | Mononucleosis - IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum |
Sample Type | Serum |
Type | Reagent |
Quantity | 100 Tests |
Unit Size | Each |
Description | ImmunoDOT™ Mono IgG |
Detectable Analytes | Mononucleosis - IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum |
Sample Type | Serum |
Type | Reagent |
Quantity | 25 Tests |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Detectable Analytes | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R246019 | Each | ImmunoDOT™ Mono IgG | Mononucleosis - IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum | 100 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R246015 | Each | ImmunoDOT™ Mono IgG | Mononucleosis - IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum | 25 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|