Remel ImmunoDOT Borrelia is an EIA membrane test used for the qualitative detection of Borrelia burgdorferi IgA, IgG, IgM and Anti-P39 antibodies for the diagnosis of Lyme disease.
Quantity | 25 Tests/Kit |
Description | ImmunoDOT™ Borrelia |
Detectable Analytes | Borrelia burgdorferi IgA, IgG, IgM, and Anti-P39 antibodies for the diagnosis of Lyme disease |
Type | Borrelia Testing Kit |
Unit Size | Each |
Quantity | 100 Tests/Kit |
Description | ImmunoDOT™ Borrelia |
Detectable Analytes | Borrelia burgdorferi IgA, IgG, IgM, and Anti-P39 antibodies for the diagnosis of Lyme disease |
Type | Borrelia Testing Kit |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Detectable Analytes | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R2470445 | Each | ImmunoDOT™ Borrelia | Borrelia burgdorferi IgA, IgG, IgM, and Anti-P39 antibodies for the diagnosis of Lyme disease | 25 Tests/Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R2470882 | Each | ImmunoDOT™ Borrelia | Borrelia burgdorferi IgA, IgG, IgM, and Anti-P39 antibodies for the diagnosis of Lyme disease | 100 Tests/Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|