Thermo Scientific™

Lysis Buffer for MagJET Plant RNA Kit

Catalog number: R2542
Thermo Scientific™

Lysis Buffer for MagJET Plant RNA Kit

Catalog number: R2542
Catalog Number
R2542
Unit Size
140 mL
Price (USD)
Price: 199.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R2542140 mL
Price: 199.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific MagJET Lysis Buffer is a component of the MagJET Plant RNA DNA Kits (Cat. No. K2771, K2772) and may be purchased separately.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10X
Buffer
Lysis Buffer
For Use With (Equipment)
MagJET Plant gDNA Kit
Product Line
MagJET
Quantity
140 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text