You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
Home
Shop All Products
Molecular Biology Reagents and Kits
RNA Isolation and Purification Products
Wash Buffer 1 for MagJET Plant RNA Kit (concentrated)
Thermo Scientific™
Wash Buffer 1 for MagJET Plant RNA Kit (concentrated)
Catalog number:
R2551
Related applications:
DNA Extraction
Technical Support
Customer Service
Thermo Scientific™
Wash Buffer 1 for MagJET Plant RNA Kit (concentrated)
Catalog number:
R2551
Related applications:
DNA Extraction
Technical Support
Customer Service
Catalog Number
R2551
Unit Size
110 ml
Price (USD)
Price:
78.50
Your Price:
Sign In
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
R2551
110 ml
Price:
78.50
Your Price:
Sign In
-
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific MagJet Wash Buffer 1 (conc.) is a component of the MagJET Plant RNA DNA Kits (K2771/2772) and may be purchased separately. Provided in 110 mL aliquots.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer
Wash Buffer
For Use With (Equipment)
MagJET Plant RNA Kit
Product Line
MagJET
Quantity
110 mL
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search