Wash Buffer 2 for MagJET Viral Kit (concentrated)
Catalog number: R2661
Thermo Scientific™  Related applications: Viral RNA⁄DNA Purification
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific MagJet Wash Buffer 2 (conc.) is a component of the MagJET Viral DNA and RNA Kits (K2781/2782) and may be purchased separately. Provided in 50 mL aliquots.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer Type: Wash Buffer
Product Line: MagJET
Product Size: 50 mL
Documents
Manuals & protocols