TopVision Agarose Tablets
Thermo Scientific™

TopVision Agarose Tablets

The Thermo Scientific TopVision Agarose Tablets, low EEO (LE), are a multi-purpose agarose in tablet format delivered in a convenientRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R28021000 Tablets (0.5 g each)
R2801200 Tablets (0.5 g each)
Catalog number R2802
Price (USD)
636.65
Online exclusive
796.00
Save 159.35
Each
Quantity:
1000 Tablets (0.5 g each)
Request bulk or custom format
Price (USD)
636.65
Online exclusive
796.00
Save 159.35
Each
The Thermo Scientific TopVision Agarose Tablets, low EEO (LE), are a multi-purpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.

Features
• Convenient tablet format—no messy weighing required
• Fast dissolving protocol
• 'Green'—manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
• Enhanced resolution and reduced background

Applications

• Ideal for routine DNA and RNA gel electrophoresis and blotting assays.

Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Melting Point88 °C
Quantity1000 Tablets (0.5 g each)
Shelf Life2 Years
Validated ApplicationGel Electrophoresis
Product LineTopVision
Product TypeTop Vision Agarose Tablet
Separation Range100 to >30000 bp
Unit SizeEach

Frequently asked questions (FAQs)

How long does it take to dissolve a Thermo Scientific TopVision Agarose Tablet?

Add the tablet into the determined amount of buffer, presoak for two minutes, and proceed the same way as you are used to with the traditional agarose powder. Using tablets saves 15-20 minutes hands-on-time.

At what voltage should I run an agarose gel prepared from TopVision Agarose Tablets?

The electrophoresis conditions do not depend on the form of delivery of agarose. Whether it is loose powder or tablets, the material has the same physicochemical conditions. In general, 4-10 volts/cm (cm is determined by measuring the interelectrode distance, not the gel length) are recommended when running agarose gels under normal horizontal electrophoretic conditions.

Do TopVision Agarose Tablets perform the same compared to traditional agarose powder products?

Yes, the performance is the same, the convenience is much greater.The critical characteristics of agaroses (melting/gelling temperature, gel strength, electroendosmosis (EEO)) impacting the performance are the same.

What is the advantage of using agarose tablets rather than loose powder agarose?

A tablet is a more convenient format, eliminating the messy and time-consuming weighing step.