Thermo Scientific™
Remel™ Agglutinating Sera, Salmonella H
Catalog number: R30160701
Thermo Scientific™
Remel™ Agglutinating Sera, Salmonella H
Catalog number: R30160701
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Catalog Number
R30160701
Unit Size
Each
Description
Salmonella H (eh)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (eh)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (i)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (i)
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella H
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H ©
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H ©
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (a)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (a)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (gq)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (gq)
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella H
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (1, 7)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (1, 7)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (enz15)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (enz15)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (G) Polyvalent, for gm, gp, fg, gq, fgt, gms, gmt, gt, gmq, gpu
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (G) Polyvalent, for gm, gp, fg, gq, fgt, gms, gmt, gt, gmq, gpu
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
MaterialPolyvalent
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (enx)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (enx)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella H (b)
Detectable Analytes
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella H (b)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Showing 10 of 39Show more
Catalog NumberSpecificationsUnit SizeDescriptionDetectable AnalytesQuantityPrice (USD)
R30160701Full specifications
EachSalmonella H (eh)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (eh)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30161601Full specifications
EachSalmonella H (i)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (i)
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella H
Unit SizeEach
R30160401Full specifications
EachSalmonella H ©flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H ©
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30160201Full specifications
EachSalmonella H (a)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (a)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30161401Full specifications
EachSalmonella H (gq)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (gq)
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella H
Unit SizeEach
R30163601Full specifications
EachSalmonella H (1, 7)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (1, 7)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30160901Full specifications
EachSalmonella H (enz15)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (enz15)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30161001Full specifications
EachSalmonella H (G) Polyvalent, for gm, gp, fg, gq, fgt, gms, gmt, gt, gmq, gpuflagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (G) Polyvalent, for gm, gp, fg, gq, fgt, gms, gmt, gt, gmq, gpu
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
MaterialPolyvalent
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30160801Full specifications
EachSalmonella H (enx)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (enx)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
R30160301Full specifications
EachSalmonella H (b)flagellar H antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella H (b)
CE MarkerYes
Detectable Analytesflagellar H antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
Unit SizeEach
Showing 10 of 39Show more
  • Select sera also for use as positive controls for stained Salmonella suspensions
Figures