Thermo Scientific™
Remel™ Agglutinating Sera, Alkalescens-Dispar
Catalog number: R30164301
Thermo Scientific™
Remel™ Agglutinating Sera, Alkalescens-Dispar
Catalog number: R30164301
Remel Agglutinating Sera are used for serological identification of Shigella cultures by rapid slide agglutination test.
Catalog Number
R30164301
Unit Size
Each
Quantity
2 mL
Price (USD)
Full specifications
DescriptionAlkalescens-Dispar Agglutinating Sera
FormatVial
Packaging Type2 mL/Vial
Quantity2 mL
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R30164301Full specifications
Each2 mLRequest A Quote
DescriptionAlkalescens-Dispar Agglutinating Sera
FormatVial
Packaging Type2 mL/Vial
Quantity2 mL
Unit SizeEach
Showing 1 of 1
Remel Agglutinating Sera are used for serological identification of Shigella cultures by rapid slide agglutination test.
Figures