Thrombo-Wellcotest™ Rapid Latex Agglutination Test
Thermo Scientific™
Thrombo-Wellcotest™ Rapid Latex Agglutination Test
Rapidly detect fibrin and fibrinogen degradation products in human serum and urine by latex agglutination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityType
R30853001100 Tubes/Pk.Collection Tubes
R308523013 mL/VialLatex
R3085260120 Tests/KitKit
R30853101100 mL/BottleGlycine Saline Buffer
Catalog number R30853001
Price (MXN)
-
Quantity:
100 Tubes/Pk.
Type:
Collection Tubes
Carry out semi-quantitative detection of fibrin and fibrinogen degradation products in human serum and urine using Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test.
Use this simple, rapid, semi-quantitative test for the presence of fibrin/fibrinogen degredation products (FDP) in human serum and urine samples.
The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.
Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.
The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:
  • Thrombo-Wellcotest Latex (Part No. R30852301)
  • Thrombo-Wellcotest Collection Tubes (Part No. R30853001)—Glass tubes contain soya bean trypsin inhibitor (approximately 3600 NF units/tube) and Bothrops atrox venom (>10μg/tube) for the collection of 2mL whole blood or urine.
  • Thrombo-Wellcotest Glycine Saline Buffer (Part No. R30853101)—Glycine saline buffer is provided as a replacement item in instances where multiple tests have exhausted the supply of this component.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Quantity100 Tubes/Pk.
TypeCollection Tubes
Unit SizeEach
Contents & Storage
2°C to 8°C