Wellcogen™ Haemophilus influenzae b Rapid Latex Agglutination Test
Thermo Scientific™
Wellcogen™ Haemophilus influenzae b Rapid Latex Agglutination Test
Rapidly detect antigens to Haemophilus influenzae present in human body fluids or blood cultures by latex agglutination.
Have Questions?
Número do catálogoQuantity
R3085880130 Tests
Número do catálogo R30858801
Preço (BRL)
-
Quantity:
30 Tests
Wellcogen™ H. influenzae b is a rapid latex test for use in the qualitative detection of antigen from Haemophilus influenzae type b, present in body fluids as a consequence of infection or in blood cultures. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; eg symptoms, results of other tests, clinical impressions etc
Use this rapid latex test in the qualitative detection of antigen from Haemophilus influenzae type b, present in body fluids as a consequence of infection, or in blood cultures.
The reagent consists of polystyrene latex particles which have been coated with antibodies specific to H. influenzae type b antigen. These latex particles agglutinate in the presence of sufficient homologous antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificações
DescriptionWellcogen Haemophilus influenzae b Test Kit
Quantity30 Tests
Sample TypeBody Fluids, Blood Culture
TypeReagent
Detectable AnalytesInfluenza B
Unit SizeEach
Conteúdo e armazenamento
2°C to 8°C