| R30957501 | Full specifications | Each | Salmonella O Factor 10 (group E1) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 10 (group E1) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group E1 | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30959001 | Full specifications | Each | Salmonella O Factor 38 (group P) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 38 (group P) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group P | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30958901 | Full specifications | Each | Salmonella O Factor 35 (group O) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 35 (group O) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group O | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30958801 | Full specifications | Each | Salmonella O Factor 30 (group N) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 30 (group N) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group N | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30959201 | Full specifications | Each | Salmonella O Factor 40 (group R) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Organism Group | Group R | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Description | Salmonella O Factor 40 (group R) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Unit Size | Each |
|
| R30956801 | Full specifications | Each | Salmonella O Factors 3, 10, 15, 19 (group E) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factors 3, 10, 15, 19 (group E) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group E | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30958001 | Full specifications | Each | Salmonella O Factor 16 (group I) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 16 (group I) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group I | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30958701 | Full specifications | Each | Salmonella O Factor 28 (group M) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 28 (group M) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group M | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30957601 | Full specifications | Each | Salmonella O Factor 11 (group F) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 11 (group F) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group F | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|
| R30956701 | Full specifications | Each | Salmonella O Factor 2 (group A) | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | 2 mL | Request A Quote |
Control Sets | Positive | Description | Salmonella O Factor 2 (group A) | Detectable Analytes | somatic O antigens of Salmonella strains by slide and/or tube agglutination test | Type | Agglutinating Sera | Organism Group | Group A | Packaging Type | 2 mL/Vial | Quantity | 2 mL | Target Organism Class | Salmonella O | Unit Size | Each |
|