Thermo Scientific™

Remel™ Agglutinating Sera, Salmonella O

Catalog number: R30958601
Thermo Scientific™

Remel™ Agglutinating Sera, Salmonella O

Catalog number: R30958601
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
 
Catalog Number
R30958601
Unit Size
Each
Description
Salmonella O Factor 27 (group B)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 27 (group B)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup B
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 38 (group P)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 38 (group P)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup P
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 35 (group O)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 35 (group O)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup O
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 30 (group N)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 30 (group N)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup N
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 40 (group R)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
Organism GroupGroup R
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
DescriptionSalmonella O Factor 40 (group R)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factors 3, 10, 15, 19 (group E)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factors 3, 10, 15, 19 (group E)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup E
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 16 (group I)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 16 (group I)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup I
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 28 (group M)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 28 (group M)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup M
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 11 (group F)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 11 (group F)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup F
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Salmonella O Factor 2 (group A)
Detectable Analytes
somatic O antigens of Salmonella strains by slide and/or tube agglutination test
Quantity
2 mL
Price (USD)
Full specifications
Control SetsPositive
DescriptionSalmonella O Factor 2 (group A)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup A
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Showing 10 of 26Show more
Catalog NumberSpecificationsUnit SizeDescriptionDetectable AnalytesQuantityPrice (USD)
R30958601Full specifications
EachSalmonella O Factor 27 (group B)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 27 (group B)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup B
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30959001Full specifications
EachSalmonella O Factor 38 (group P)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 38 (group P)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup P
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30958901Full specifications
EachSalmonella O Factor 35 (group O)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 35 (group O)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup O
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30958801Full specifications
EachSalmonella O Factor 30 (group N)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 30 (group N)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup N
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30959201Full specifications
EachSalmonella O Factor 40 (group R)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
Organism GroupGroup R
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
DescriptionSalmonella O Factor 40 (group R)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Unit SizeEach
R30956801Full specifications
EachSalmonella O Factors 3, 10, 15, 19 (group E)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factors 3, 10, 15, 19 (group E)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup E
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30958001Full specifications
EachSalmonella O Factor 16 (group I)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 16 (group I)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup I
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30958701Full specifications
EachSalmonella O Factor 28 (group M)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 28 (group M)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup M
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30957601Full specifications
EachSalmonella O Factor 11 (group F)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 11 (group F)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup F
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
R30956701Full specifications
EachSalmonella O Factor 2 (group A)somatic O antigens of Salmonella strains by slide and/or tube agglutination test2 mLRequest A Quote
Control SetsPositive
DescriptionSalmonella O Factor 2 (group A)
Detectable Analytessomatic O antigens of Salmonella strains by slide and/or tube agglutination test
TypeAgglutinating Sera
Organism GroupGroup A
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
Unit SizeEach
Showing 10 of 26Show more
  • Select sera also for use as positive controls for stained Salmonella suspensions

Figures