Agglutinating Sera, Salmonella O
Actual product may vary
Thermo Scientific™

Agglutinating Sera, Salmonella O

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R30958901Factor 35 (group O)
R30959001Factor 38 (group P)
R30958801Factor 30 (group N)
R30959201Factor 40 (group R)
R30956801Factors 3, 10, 15, 19 (group E)
R30958701Factor 28 (group M)
R30957601Factor 11 (group F)
R30956701Factor 2 (group A)
R30959301Factor 41 (group S)
R30957101Factor 6, 7 (group C1)
R30958401Factor 20 (group C2)
R30958201Factor 18 (group K)
R30959101Factor 39 (group Q)
R30957201Factor 8 (group C2)
R30958501Factor 21 (group L)
R30957801Factor 14 (group H)
R30957001Factor 5 (group B)
R30958101Factor 17 (group J)
R30957301Factor 9 (group D)
R30956901Factor 4 (group B )
Catalog number R30958901
Price (USD)
-
Description:
Factor 35 (group O)

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.

Serologically identify somatic O antigens of Salmonella strains by slide and/or tube agglutination tests for epidemiological and diagnostic purposes using Agglutinating Sera, Salmonella O.

  • Rapid: Results within 1 minute.
  • Simple, ready to use: Makes the procedure easy to implement and run

The genus Salmonella is classified in the Kauffmann-White Scheme into serotypes according to combinations of somatic (O) and flagellar (H) antigens, which are identified by agglutination tests1. Agglutinating sera should be used in confirmatory tests but may also be used with appropriate caution in screening tests2. These simple tests rapidly detect specific bacterial antigens for the identification, grouping and/or typing of Salmonella and other food-borne microorganisms. This incorporates agglutinating sera for the identification somatic O antigens of Salmonella (groups A-S), allowing the fast, accurate and reliable identification of Salmonella strains.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Edwards, P.R. and Ewing, W.H. (1986). Identification of Enterobacteriaceae, 4th Edition, Elsevier Science Publishing Co. Inc., New York.
  2. Taylor, J. (1967). The isolation and identification of salmonellae. A.C.P. Broadsheet No. 58.
Specifications
Control SetsPositive
DescriptionFactor 35 (group O)
Organism GroupGroup O
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
TypeAgglutinating Sera
Detectable AnalytesSalmonella O Antigen
Unit SizeEach