Remel™ Wellcolex™ E. coli O157:H7
Thermo Scientific™
Remel™ Wellcolex™ E. coli O157:H7
Remel Wellcolex and E. coli O157:H7 is a rapid latex agglutination test used for presumptive identification of E. coli O157:H7 isolates.
Have Questions?
Catalog NumberQuantity
R3095960150 Tests/Kit
Catalog number R30959601
Price (MXN)
-
Quantity:
50 Tests/Kit
Remel Wellcolex and E. coli O157:H7 is a rapid latex agglutination test used for presumptive identification of E. coli O157:H7 isolates.
  • Colorimetric with red O157 latex and blue H7 latex
Specifications
DescriptionWellcolex E. coli O157:H7
Detectable AnalytesE. coli O157:H7 isolates
TypeE. coli Testing Kit
Quantity50 Tests/Kit
Unit SizeEach