Rhinohide™ Polyacrylamide Gel Strengthener Concentrate - FAQs
Rhinohide™ Polyacrylamide Gel Strengthener Concentrate - FAQs
View additional product information for Rhinohide™ Polyacrylamide Gel Strengthener Concentrate - FAQs (R33400)
1 product FAQs found
I am staining my protein gel with multiple stains over several days. How do I prevent excessive gel tearing?
For gels that you are pouring yourself, add Rhinohide Polyacrylamide Gel Strengthener (Cat. Nos. R33400, R33410) to the formulation. Decrease the shaker speed/motion; only very gentle rocking is required. Slowly decant solutions during solution changes and avoid touching the gel as much as possible when pouring out solutions.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.