Invitrogen™

RediPlate™ 96 EnzChek™ Serine/Threonine Phosphatase Assay Kit

Catalog number: R33700
Invitrogen™

RediPlate™ 96 EnzChek™ Serine/Threonine Phosphatase Assay Kit

Catalog number: R33700
Catalog Number
R33700
Unit Size
1 x 96 well plate
Price (USD)
394.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R337001 x 96 well plate
394.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
The RediPlate™ 96 EnzChek® Serine/Threonine Phosphatase Assay Kit provides a fast, simple and direct fluorescence-based assay for detecting serine/threonine phosphatases and their corresponding modulators and inhibitors. Unlike other microplate assays, this kit provides the necessary reagents predispensed into a 96-well microplate. Simply reconstitute the fluorogenic substrate in the assay wells with buffer, add the desired sample to the wells, incubate and then quantitate the fluorescence in any standard fluorescence-based microplate reader. Inhibitors are included in each assay well to ensure that the assay is selective for Ser/Thr PPases- other phosphatases, including tyrosine phosphatase do not significantly react with the substrate. Additional advantages of this assay include compatibility with non-ionic detergents and insensitivity to free phosphate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
Dye Type
DiFMU
Emission
358⁄452
Product Line
EnzChek™, RediPlate™
Shipping Condition
Room Temperature
Substrate
DiFMUP
Substrate Properties
Chemical Substrate
Substrate Type
Phosphatase Substrate
Target Enzyme
Serine⁄Threonine Phosphatases
For Use With (Equipment)
Microplate Reader
Product Type
Phosphatase Assay

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

Frequently asked questions (FAQs)

Citations & References