Propidium Iodide Ready Flow™ Reagent
Invitrogen™
Propidium Iodide Ready Flow™ Reagent
Propidium Iodide Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells inRead more
Have Questions?
Catalog NumberQuantity
R37169120 assays
Catalog number R37169
Price (USD)
144.00
Each
Quantity:
120 assays
Price (USD)
144.00
Each
Propidium Iodide Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays. This dye has excitation/emission maxima of 535/617 nm.
• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh. or pipette
• Rapid staining of dead cells without wash steps
Propidium Iodide Ready Flow Reagent is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.
Propidium iodide is membrane impermeant and therefore excluded from viable cells. It binds to DNA by intercalating between the bases, thereby binding both DNA and RNA. Propidium iodide is commonly excited with the 488 nm laser, but others such as the 532 and 561 nm lasers are also suited for PI excitation.
Specifications
Cell PermeabilityCell-Impermeant
Detection MethodFluorescence
Excitation/Emission535/617
Quantity120 assays
Reagent TypeNucleic Acid Stain
Sub Cellular LocalizationNucleus
ColorRed
Product LineReady Flow
Product TypeFluorescent Stain
Unit SizeEach
Contents & Storage
2 dropper bottles, store (at room temperature)
Frequently asked questions (FAQs)
I would like to label only 100 µL of sample of the same cell density using a Ready Flow product. The manual notes 2 drops per 1 mL (1 X 10E6 cells/mL). May I scale down for smaller volumes?
Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the concentration of dye/reagent in the Ready Flow products?
The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.