TO-PRO™3 Ready Flow™ Reagent
Invitrogen™

TO-PRO™3 Ready Flow™ Reagent

TO-PRO™3 Ready Flow™試薬は、フローサイトメトリーアッセイで生細胞と死細胞を区別するための、明るく使いやすい核酸染色試薬です。この色素の最大励起/発光波長は642/661 nmで、Alexa Fluor™ 647やアロフィコシアニン詳細を見る
製品番号(カタログ番号)数量
R37170120 assays
製品番号(カタログ番号) R37170
価格(JPY)
36,600
キャンペーン価格
Ends: 25-Dec-2026
61,000
割引額 24,400 (40%)
Each
数量:
120 assays
TO-PRO™3 Ready Flow™試薬は、フローサイトメトリーアッセイで生細胞と死細胞を区別するための、明るく使いやすい核酸染色試薬です。この色素の最大励起/発光波長は642/661 nmで、Alexa Fluor™ 647やアロフィコシアニン(APC)と同様です。

•室温で安定しています
• 便利なすぐに使えるフォーマット — 希釈、計量、ピペットでの分注が不要です
• 洗浄ステップなしで死細胞を迅速に染色します

TO-PRO 3 Ready Flow試薬は、すぐに使える、サンプルに直接使用可能な死細胞染色剤です。DNAに結合したときに赤色蛍光を発します。ドロッパーボトルから直接、1 x 106個の細胞に2滴加え、インキュベートして分析します。

TO-PRO 3 Ready Flow試薬は膜非透過性であるため、生細胞から除外されます。この試薬は選択的に二本鎖DNAに結合します。色素の最大励起/発光波長は642/661 nmです。

仕様
細胞透過性Impermeant
概要TO-PRO™3 Ready Flow™ Reagent
検出法蛍光
染色剤タイプ細胞非透過性
放射波長642/661
励起波長642 nm
形状溶液
フォーマットBottle(s)
数量120 assays
溶解性DMSO(ジメチルスルホキシド)
細胞内局在核
色遠赤色
使用対象(アプリケーション)Viability Assay
使用対象 (装置)Flow Cytometer
製品ラインReady Flow™、TO-PRO™3
製品タイプ試薬
Unit SizeEach
組成および保存条件
ドロッパーボトル2本、室温で保管してください

よくあるご質問(FAQ)

I would like to label only 100 µL of sample of the same cell density using a Ready Flow product. The manual notes 2 drops per 1 mL (1 X 10E6 cells/mL). May I scale down for smaller volumes?

Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the concentration of dye/reagent in the Ready Flow products?

The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

引用および参考文献 (1)

引用および参考文献
Abstract
Pigment epithelium-derived factor hinders photoreceptor cell death by reducing intracellular calcium in the degenerating retina.
Authors:Comitato A, Subramanian P, Turchiano G, Montanari M, Becerra SP, Marigo V,
Journal:Cell Death Dis
PubMed ID:29752430
Calcium ions play a critical role in neuronal cell death. Pigment epithelium-derived factor (PEDF) is a promising neuroprotective protein for photoreceptor cells but the mechanisms mediating its effects against retinal degeneration are still not well characterized. We addressed this question in the rd1 degenerating mouse retina that bears a mutation ... More