ReadyProbes™ Cell Viability Imaging Kit, Blue/Green
Invitrogen™

ReadyProbes™ Cell Viability Imaging Kit, Blue/Green

ReadyProbes® Cell Viability Imaging Kit (Blue/Green) is a ready-to-use assay that can quickly and easily determine the viability of cells.Read more
Have Questions?
Catalog NumberQuantity
R376091 kit
Catalog number R37609
Price (USD)
350.14
Special offer
Online exclusive
Ends: 31-Dec-2026
427.00
Save 76.86
Each
Quantity:
1 kit
Price (USD)
350.14
Special offer
Online exclusive
Ends: 31-Dec-2026
427.00
Save 76.86
Each
ReadyProbes® Cell Viability Imaging Kit (Blue/Green) is a ready-to-use assay that can quickly and easily determine the viability of cells. Just add 2 drops each of room temperature, stable NucBlue® Live reagent (Hoechst 33342) and NucGreen® Dead reagent to 1 mL of cell growth media, then determine viability by counting total vs dead cells. NucBlue® Live reagent stains the nuclei of all the cells and can be detected with a standard DAPI filter. NucGreen® Dead reagent stains only the nuclei of cells with compromised plasma membrane integrity and is detected using a standard FITC/GFP (green) filter set. This kit is appropriate for fluorescence microscopy, fluorescence microplate readers, and flow cytometry.

NucBlue® Live reagent: stains the nuclei of all cells; detected with a standard DAPI filter (excitation/emission maxima: 360/460 nm)
NucGreen® Dead reagent: stains only the nuclei of dead cells with compromised plasma membranes; detected with standard FITC/GFP (green) filter set (excitation/emission maxima: 504/523 nm)
See other ReadyProbes® reagents for cell staining
Learn more about other assays for cell viability

Suggestions for use
• NucBlue® Live and NucGreen® Dead reagents may be added directly to cells in full growth media or a compatible buffer solution.
• In most cases, 2 drops/mL and an incubation time of 5 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells, Eukaryotic Cells
DescriptionReadyProbes Cell Viability Imaging Kit, Blue/Green
Detection MethodFluorescence
Dye Type3 x 2.5 mL NucBlue™ Live
Emission Wavelength360/504
Excitation Wavelength460, 523 nm
FormatDropper bottles
Quantity1 kit
Shipping ConditionRoom Temperature
ColorBlue, Green
For Use With (Application)Viability Assay
For Use With (Equipment)Fluorescence Microscope
Product LineReadyProbes
Product TypeCell Viability Imaging Kit
Unit SizeEach
Contents & Storage
Store at 2° to 8°C, or at room temperature.

Frequently asked questions (FAQs)

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What fluorescent viability assays can I use on the Countess II FL automated cell counter?

We have validated the following kits for use on the Countess II FL Automated Cell Counter:

LIVE/DEAD Viabilty/Cytoxicity Kit (Cat. No. L3224) containing calcein AM and ethidium homodimer-1
ReadyProbes Cell Viability Imaging Kit, Blue/Green (Cat. No. R37609)
ReadyProbes Cell Viability Imaging Kit, Blue/Red (Cat. No. R37610) containing NucBlue Live/NucGreen Dead and NucBlue Live/propidium iodide
See this Application Note for details - https://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/Laboratory%20Instruments/Files/0415/CO014723-Countess-II-FL-Viability-Appnote_FHR.pdf.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between NucBlue Live reagents stain and NucGreen Dead reagents stain?

The NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (15)

Citations & References
Abstract
In vitro studies with renal proximal tubule cells show direct cytotoxicity of Androctonus australis hector scorpion venom triggered by oxidative stress, caspase activation and apoptosis.
Authors:Saidani C, Hammoudi-Triki D, Laraba-Djebari F, Taub M,
Journal:Toxicon
PubMed ID:27470530
'Scorpion envenomation injures a number of organs, including the kidney. Mechanisms proposed to explain the renal tubule injury include direct effects of venom on tubule epithelial cells, as well as indirect effects of the autonomic nervous system, and inflammation. Here, we report direct effects of Androctonus australis hector (Aah) scorpion ... More
Fluorescence and Cytotoxicity of Cadmium Sulfide Quantum Dots Stabilized on Clay Nanotubes.
Authors:Stavitskaya AV, Novikov AA, Kotelev MS, Kopitsyn DS, Rozhina EV, Ishmukhametov IR, Fakhrullin RF, Ivanov EV, Lvov YM, Vinokurov VA
Journal:Nanomaterials (Basel)
PubMed ID:29857546
'Quantum dots (QD) are widely used for cellular labeling due to enhanced brightness, resistance to photobleaching, and multicolor light emissions. CdS and Cd'
Chemobrionic structures in tissue engineering: self-assembling calcium phosphate tubes as cellular scaffolds.
Authors:Hughes EAB, Chipara M, Hall TJ, Williams RL, Grover LM
Journal:Biomater Sci
PubMed ID:31830151
'A diverse range of complex patterns and mineralised hierarchical microstructures can be derived from chemobrionic systems, with formation driven by complex reaction-diffusion mechanisms far from thermodynamic equilibrium. In these experiments, self-assembling calcium phosphate tubes are generated using hydrogels made with 1 M calcium solutions layered with solutions of dibasic sodium ... More
An evolutionary perspective on the role of mesencephalic astrocyte-derived neurotrophic factor (MANF): At the crossroads of poriferan innate immune and apoptotic pathways.
Authors:Sereno D, Müller WEG, Bausen M, Elkhooly TA, Markl JS, Wiens M,
Journal:Biochem Biophys Rep
PubMed ID:28955781
'The mesencephalic astrocyte-derived neurotrophic factor (MANF) belongs to a recently discovered family of neurotrophic factors. MANF can be secreted but is generally resident within the endoplasmic reticulum (ER) in neuronal and non-neuronal cells, where it is involved in the ER stress response with pro-survival effects. Here we report the discovery ... More
A novel method to understand tumor cell invasion: integrating extracellular matrix mimicking layers in microfluidic chips by
Authors:Eslami Amirabadi H, SahebAli S, Frimat JP, Luttge R, den Toonder JMJ,
Journal:Biomed Microdevices
PubMed ID:29038872
'A major challenge in studying tumor cell invasion into its surrounding tissue is to identify the contribution of individual factors in the tumor microenvironment (TME) to the process. One of the important elements of the TME is the fibrous extracellular matrix (ECM) which is known to influence cancer cell invasion, ... More