ReadyProbes™ 2.5% Normal Goat Serum (1X)
Invitrogen™

ReadyProbes™ 2.5% Normal Goat Serum (1X)

ReadyProbes 2.5% Normal Goat Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF)Read more
Have Questions?
Catalog NumberQuantity
R3762450 mL
Catalog number R37624
Price (USD)
145.14
Special offer
Online exclusive
Ends: 31-Dec-2026
177.00
Save 31.86
Each
Quantity:
50 mL
Price (USD)
145.14
Special offer
Online exclusive
Ends: 31-Dec-2026
177.00
Save 31.86
Each
ReadyProbes 2.5% Normal Goat Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments, usually when a goat secondary antibody is used. Incubating cells/tissue with diluted serum from the same species as the host of the secondary antibody typically reduces non-specific binding. Simply cover the tissue section or cells with a few drops of ReadyProbes 2.5% Normal Goat Serum and incubate for 30–60 minutes at room temperature. Ready Probes 2.5% Normal Goat Serum can also be used to dilute primary and secondary antibodies.

Ready Probes 2.5% Normal Goat Serum is provided as a ready-to-use reagent with no further dilution or preparation required. The serum is collected from healthy adult animals and the heat-treated, centrifuged, and filtered. It is then checked for cross reactivity using an appropriate immunoassay. The serum is diluted to 2.5% (v/v) in buffer, with 0.08% sodium azide added as a preservative.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineReadyProbes
Reagent TypeBlocking Reagent
Shipping ConditionRoom Temperature or Wet Ice
SpeciesGoat
FormLiquid
Product TypeGoat Serum
Quantity50 mL
Unit SizeEach
Contents & Storage
Provided as 50-mL solution at ready-to-use (1X) concentration in a clear plastic dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Frequently asked questions (FAQs)

Can I use ReadyProbes 2.5% Normal Goat Serum (1X) as an antibody diluent for both primary and secondary antibodies?

Yes, ReadyProbes 2.5% Normal Goat Serum (1X) can be used as an antibody diluent for both primary and secondary antibodies, except for HRP-conjugated antibodies. The sodium azide added as a preservative is an inhibitor of horseradish peroxidase.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How many drops of ReadyProbes 2.5% Normal Goat Serum (1X) should I add to a sample?

Add a sufficient number of drops of ReadyProbes 2.5% Normal Goat Serum (1X) to completely cover the sample.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When would I use goat, horse, or chicken serum for blocking versus FBS or BSA?

For blocking, you should use a serum of the same or closely related species of the secondary antibody being used. If using a secondary antibody that was raised in goat or sheep, use goat serum (Cat. No. R37624 or 31873) for blocking; if using a secondary antibody raised in donkey or horse, use horse serum (Cat. No. R37625 or 31874) for blocking; if using a secondary antibody raised in chicken, use chicken serum (Cat. No. R37626) for blocking. FBS and BSA are alternatives when a specific species of serum is not available.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.