Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
Volume (Metric) | 5 x 250 mL |
Type | Enhancer |
Quantity | 250mL/Bottle |
Unit Size | Each |
Volume (Metric) | 3.785 L |
Type | Enhancer |
Quantity | Each |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R40231 | Each | Enhancer | 250mL/Bottle | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R40232 | Each | Enhancer | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.