Product Image
Thermo Scientific™
Remel™ SAS™ Adeno Test
Cassette immunoassay used for qualitative detection of adenovirus antigens in eye swabs, nasal and pharyngeal secretions, and fecal material.
Have Questions?
Catalog NumberQuantity
R4206702020 Tests/Kit
Catalog number R42067020
Price (CLP)
-
Quantity:
20 Tests/Kit
Thermo Scientific™ Remel SAS Adeno Test is a cassette immunoassay used for qualitative detection of adenovirus antigens in eye swabs, nasal and pharyngeal secretions, and fecal material.
SA Scientific
Specifications
DescriptionSAS Adeno Test
Quantity20 Tests/Kit
Detectable AnalytesAdenovirus antigen
Unit SizeEach