Thermo Scientific™

Remel™ Egg Yolk Suspension 50%

Catalog number: R450290
Thermo Scientific™

Remel™ Egg Yolk Suspension 50%

Catalog number: R450290

Remel Egg Yolk Suspension 50% is used with McClung-Toabe Agar Base.

 
Catalog Number
R450290
Unit Size
Each
Quantity
100 mL
Price (USD)
Full specifications
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Suspension 50%
Product TypeSuspension
Quantity100 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
100 mL
Price (USD)
Full specifications
DescriptionEgg Yolk Suspension 50%
Product TypeSuspension
Quantity100 mL
TypeSupplement
FormatLiquid
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R450290Full specifications
Each100 mLRequest A Quote
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Suspension 50%
Product TypeSuspension
Quantity100 mL
Unit SizeEach
R450291Full specifications
Each100 mLRequest A Quote
DescriptionEgg Yolk Suspension 50%
Product TypeSuspension
Quantity100 mL
TypeSupplement
FormatLiquid
Unit SizeEach
Showing 2 of 2

Remel Egg Yolk Suspension 50% is used with McClung-Toabe Agar Base.

Figures

Frequently asked questions (FAQs)

Citations & References