Thermo Scientific™

Remel™ Egg Yolk Tellurite 100%

Catalog number: R450332
Thermo Scientific™

Remel™ Egg Yolk Tellurite 100%

Catalog number: R450332

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

 
Catalog Number
R450332
Unit Size
Each
Packaging Type
10/Pk.
Quantity
10 mL
Price (USD)
Full specifications
Packaging Type10/Pk.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity10 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Packaging Type
1 Ea.
Quantity
100 mL
Price (USD)
Full specifications
Packaging Type1 Ea.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity100 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Packaging Type
10/Pk.
Quantity
100 mL
Price (USD)
Full specifications
Packaging Type10/Pk.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity100 mL
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizePackaging TypeQuantityPrice (USD)
R450332Full specifications
Each10/Pk.10 mLRequest A Quote
Packaging Type10/Pk.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity10 mL
Unit SizeEach
R450330Full specifications
Each1 Ea.100 mLRequest A Quote
Packaging Type1 Ea.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity100 mL
Unit SizeEach
R450331Full specifications
Each10/Pk.100 mLRequest A Quote
Packaging Type10/Pk.
FormatLiquid
TypeSupplement
DescriptionEgg Yolk Tellurite 100%
Product TypeTellurite
Quantity100 mL
Unit SizeEach
Showing 3 of 3

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

Figures

Frequently asked questions (FAQs)

Citations & References