Burkholderia cepacia Sel Agar (USP 60) 500g
Thermo Scientific™

Burkholderia cepacia Sel Agar (USP 60) 500g

Compliance to the USP 60 regulations for B. cepacia complex made simple
Have Questions?
Catalog number R452642
Price (USD)
-
Burkholderia cepacia medium (USP 60) is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in products, especially those for inhalation use of non-sterile acqueous preparations for oral, oromucosal, cutaneous or nasal use.

Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.

  • Compliant formulation according to USP 60
  • Quality tested to meet USP 60 specifically for Pharma customers
  • Part of a complete workflow which include DCM and QCO
For Laboratory Use
For Laboratory Use
Specifications
DescriptionBurkholderia cepacia Selective agar (USP 60)
For Use With (Application)Pharmaceutical testing of non-sterile products used for nasal inhalation
Format500g Pot
Quantity500 g
Regulatory StatusLab use only
Product TypeAgar
Unit SizeEach