Search
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other gram-negative, non-fermentative bacteria using this medium.
| Catalog Number | Quantity |
|---|---|
| R452802 | 500 g |
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other non-fermentative, gram-negative bacilli using Thermo Scientific™ Remel™ Cetrimide Selective Agar (Dehydrated). King et al. developed a medium called Tech Agar which was used to enhance pyocyanin production by Pseudomonas1. In 1955, Lowbury described the use of cetrimide in a selective medium for P. aeruginosa2. This formulation is recommended by the United States Pharmacopeia (USP) and the AOAC International (AOAC)3,4. Centrimide Agar has the formula of Tech Agar but is modified by the addition of centrimide.
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other non-fermentative, gram-negative bacilli using Cetrimide Selective Agar (Dehydrated).
This medium contains potassium sulfate and magnesium chloride which stimulate the production of pyocyanin and fluorescein. Fluorescein production is visualized using an ultraviolet light. P. aeruginosa is characterized by production of pyocyanin, a blue-green, water-soluble, non-fluorescent phenazine pigment. Centrimide (N-acetyl-NNN-trimethyl-ammonium bromide, cetavlon) causes nitrogen and phosphorus to be released from bacterial cells and inhibits bacteria other than P. aeruginosa.