Thermo Scientific™ Remel Modified EC Medium is used with novobiocin for the selective isolation of Escherichia coli O157:H7.
Thermo Scientific™ Remel Modified EC Medium is used with novobiocin for the selective isolation of Escherichia coli O157:H7.