Product Image
Thermo Scientific™
Remel™ McClung-Toabe Agar Base
Isolate and differentiate Clostridium spp. using McClung-Toabe Agar (Dehydrated) and Egg yolk suspension (50%).
Have Questions?
카탈로그 번호수량
R453932500 g
카탈로그 번호 R453932
제품 가격(KRW)
-
수량:
500 g
Isolate and differentiate Clostridium spp. using Thermo Scientific™ Remel™ McClung-Toabe Agar (Anaerobic Lecithin-Lipase Agar) (Dehydrated) supplemented with Egg yolk suspension (50%) R450290 or R450291. In 1947, McClung and Toabe developed this differential plating medium to detect lecithinase and lipase production by Clostridium spp1. This agar was further modified by Dowell and Hawkins to selectively isolate and differentiate certain obligately anaerobic bacteria2.
Use McClung-Toabe Agar to isolate and differentiate Clostridium spp.
Clostridium spp. produce the enzymes lecithinase and lipase. Egg yolk suspension (50%) added to the medium serves as the substrate to detect lecithinase and lipase activity3. Lecithinase degrades lecithin and produces an insoluble, opaque precipitate in the medium surrounding growth4. Lipase breaks down free fats in the egg yolk forming in an iridescent sheen on the colony surface.
This medium contains gelatin peptone that supplies amino acids and other nitrogenous compounds necessary for the growth of anaerobic bacteria. Dextrose serves as an energy source. Egg yolk serves as a substrate for detection of lecithinase and lipase activity3 and reduces the toxic effect of organic peroxides which may accumulate in the medium. Agar is the solidifying agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
  1. McClung, L.S. and R. Toabe. 1947. J. Bacteriol. 53:139-147.
  2. Dowell, V.R. and T.M. Hawkins. 1977. Laboratory Methods in Anaerobic Microbiology, CDC, Atlanta, GA.
  3. Murray, P.R., E.J. Baron, J.H. Jorgensen, M.L. Landry, and M.A. Pfaller. 2007. Manual of Clinical Microbiology. 9th ASM Press, Washington, D.C.
  4. Dowell, V.R., G.L. Lombard, F.S. Thompson, and A.Y. Armfield. 1987. Media for the Isolation, Characterization, and Identification of Obligately Anaerobic Bacteria. CDC. Atlanta, GA.
사양
설명McClung Toabe Agar Base
수량500 g
형태Powder
제품 유형Agar
Unit SizeEach