Product Image
Thermo Scientific™
Mueller Hinton Agar
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R4540842.5 kg
R454082500 g
R45408610 kg
Catalog number R454084
Price (MXN)
-
Quantity:
2.5 kg
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Specifications
Certifications/ComplianceConforms to CLSI standards. Industrial Reference: BAM, COMPF.
DescriptionMueller Hinton Agar
Quantity2.5 kg
FormPowder
Product TypeAgar
Unit SizeEach