This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Acinetobacter sp. ATCC™ 49139™
Thermo Scientific™
Culti-Loops™ Acinetobacter sp. ATCC™ 49139™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46005015 Loops/Pack
Catalog number R4600501
Price (USD)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®49139TM*
DescriptionAcinetobacter sp.
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsAmMS 202
Product TypeCulti-Loops Quality Control Organism
SpeciesAcinetobacter species
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Unit SizeEach