This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Enterococcus faecalis ATCC™ 33186™
Thermo Scientific™

Culti-Loops™ Enterococcus faecalis ATCC™ 33186™

Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.
Have Questions?
Catalog NumberQuantity
R46013015 Loops/Pk.
Catalog number R4601301
Price (USD)
-
Quantity:
5 Loops/Pk.

Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.

Enterococcus faecalis™ ATCC™ 33186™ is recommended to be used to assess suitability of Mueller Hinton Agar or broth for sulfonamide and trimethoprim MIC & disc diffusion tests (eg. trimethoprim-sulfamethoxazole discs).

Enterococcus faecalis™ ATCC™ 33186™ can be used to assess Mueller-Hinton medium suitability for sulfonamide and trimethoprim testing.

Mueller-Hinton agar or broth medium containing excessive amounts of thymidine or thymine can reverse the inhibitory effect of sulfonamides and trimethoprim, which may result in smaller and less distinct disc diffusion zones and false-resistance reports.

With broth microdilution MIC testing, suitability of Mueller-Hinton broth for sulfonamide & trimethoprim tests can be evaluated by reviewing ease of reading the results: the end points should be clear at 80% or greater reduction in growth compared to the control.

Technique:

The loop can be streaked directly onto appropriate media.

Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.

Includes:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ATCC®ATCC™ 33186™
DescriptionEnterococcus faecalis™ ATCC™ 33186™
FormatCulti-Loops
Quantity5 Loops/Pk.
Strain DesignationsCN 478
Target Organism ClassGram-positive Cocci
Test TypeQualitative QC
Unit SizeEach