This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Streptococcus agalactiae group B ATCC™ 12386™
Thermo Scientific™
Culti-Loops™ Streptococcus agalactiae group B ATCC™ 12386™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46070275 Loops/Pack
Catalog number R4607027
Price (USD)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®12386TM*
DescriptionStreptococcus agalactiae group B ATCC™ 12386™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain Designationsgrouping strain O90R [E. Todd strain Aronson Wamoscher]
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassCocci
Test TypeQualitative QC
Unit SizeEach