This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Streptomyces griseus subsp. griseus ATCC™ 10137™
Thermo Scientific™
Culti-Loops™ Streptomyces griseus subsp. griseus ATCC™ 10137™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46090135 Loops/Pack
Catalog number R4609013
Price (KRW)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®10137TM*
DescriptionStreptomyces griseus subsp. griseus ATCC™ 10137™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain Designations4 [BUCSAV 7, Boots FD40, NCIB 8232, NCTC 6961]
Product TypeCulti-Loop
Target Organism ClassGram-positive Rods
Test TypeQualitative QC
Unit SizeEach