Culti-Loops™ Hanseniaspora valbyensis ATCC™ 58370™
Thermo Scientific™
Culti-Loops™ Hanseniaspora valbyensis ATCC™ 58370™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46093985 Loops
Catalog number R4609398
Price (CLP)
-
Quantity:
5 Loops
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®58370TM*
DescriptionHanseniaspora valbyensis ATCC™ 58370™
FormatCulti-Loops
Packaging Type5 Loops/Pack
Quantity5 Loops
Strain DesignationsNRRL Y1382 [CBS 281]
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassFungi
Test TypeQualitative QC
Unit SizeEach