This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops&trade; <i>Campylobacter jejuni</i> ATCC&reg; 33560&trade;
Thermo Scientific™

Culti-Loops™ Campylobacter jejuni ATCC® 33560™

Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.
Have Questions?
Catalog NumberQuantity
R46094985 Loops/Pk
Catalog number R4609498
Price (USD)
-
Quantity:
5 Loops/Pk

Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.

Culti-Loops™ Campylobacter jejuni ATCC® 33560™ is recommended to be used for antimicrobial susceptibility quality control testing.

Campylobacter jejuni ATCC® 33560™ is recommended to be used for antimicrobial susceptibility quality control testing by CLSI and EUCAST.

Technique:

The loop may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000. Alternatively, it can be streaked directly onto appropriate media.

Includes:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

Specifications
ATCC®ATCC™ 33560™
DescriptionCampylobacter jejuni ATCC™ 33560™
FormatCulti-Loops
Quantity5 Loops/Pk
Strain DesignationsNCTC 11351, CIP 70.2T, DSM 4688, CCUG 11284
Target Organism ClassGram-negative, Helical-shaped bacteria
Test TypeQualitative QC
Unit SizeEach