Thermo Scientific™
Culti-Loops™ Coagulase Test QC Set
Related applications:
Clinical Microbiology
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Back to top
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Staphylococcus aureus subsp. aureus ATCC™ 25923™†
Staphylococcus epidermidis ATCC™ 12228™†
Ordering Information:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.