This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Vitek™ 2 QC Sets
Vitek™ 2 QC Sets
Vitek™ 2 QC Sets
Thermo Scientific™
Vitek™ 2 QC Sets
Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
R46520225 Loops/Pk.YST Comp QC Set
R46520185 Loops/Pk.AST-GN QC Set
R46520205 Loops/Pk.GP Comp QC Set
R46520215 Loops/Pk.GN Comp QC Set
R46520195 Loops/Pk.AST-GP QC Set
R46520245 Loops/Pk.NH Comp QC Set
R46520305 Loops/Pk.ANC Comp QC Set
R46820215 Loops/Pk.GN Comp QC 1 Loop Set
R46820305 Loops/Pk.ANC Comp QC 1 Loop Set
R46820184 Loops/SetAST-GN QC 1 Loop Set
R46820195 Loops/Pk.AST-GP QC 1 Loop Set
R46820205 Loops/Pk.GP Comp QC 1 Loop Set
Catalog number R4652022
Price (USD)
-
Quantity:
5 Loops/Pk.
Product Type:
YST Comp QC Set
Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ Vitek™ 2 QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.
Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.
Convenience
  • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.
Performance
  • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.
Value
  • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.
Confidence
  • Reach complete traceability with ATCC™ License Derivative strains.
Efficiency
  • Achieve accurate and convenient documentation with peel-off, transferable labels.
Safety
  • Reduce the risk of contamination and infection with zero organism handling.
  • More than 600 quality control organisms, including convenient sets for commonly used identification systems
  • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
  • ATCC™ Licensed Derivative Program member
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC™. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
ATCC®33389TM*, MYA-2950™*, 34499™*, 9950™*, 58370™*, 43534™*, 16529™*, MYA-4550™*, 12228™*, 204094™*, MYA-4549™*
Certifications/ComplianceISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
DescriptionVitek 2 YST Comp QC Set/11
FormatCulti-Loops
Quantity5 Loops/Pk.
Product TypeYST Comp QC Set
SpeciesCandida alibicans, Candida glabrata, Candida lusitaniae, Candida utilis, Kloeckera japonica, Oligella ureolytica, Prototheca wickerhamii, Sporidiobolus salmonicolor, Staphylococcus epidermidis, Trichosporon mucoides, Zygosaccharomyces bailii
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach
Contents & Storage
2°C to 8°C