Search
| 카탈로그 번호 | 설명 |
|---|---|
| R4653061 | API Rapid 20E QC Set/3 |
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedure.
Includes:
Escherichia coli ATCC™ 11775™†
Klebsiella pneumoniae subsp.pneumoniae ATCC™ 35657™†
Proteus hauseri ATCC™ 13315™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.