Culti-Loops™ API™ Staph QC Set
Thermo Scientific™
Culti-Loops™ API™ Staph QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Número do catálogoQuantity
R46530654 Loops/Pack
Número do catálogo R4653065
Preço (BRL)
-
Quantity:
4 Loops/Pack
Thermo Scientific™ Culti-Loops™ API Staph Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Staphylococcus capitis subsp. capitis ATCC™ 35661™†
Staphylococcus lentus ATCC™ 700403™†
Staphylococcus xylosus ATCC™ 700404™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Especificações
ATCC®35661TM*
DescriptionAPI Staph QC Set/3
FormatCulti-Loops
Quantity4 Loops/Pack
Product TypeCulti-Loops QC Set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach