Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Thermo Scientific™
Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberProduct Type
R4718001Quality Control Kit
Catalog number R4718001
Price (USD)
-
Product Type:
Quality Control Kit
Thermo Scientific™ Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™ is supplied as a film of preserved microorganisms inside the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®8100TM*
DescriptionSerratia marcescens ATCC™ 8100™
FormatQuanti-Cult Plus
Strain DesignationsNRS 116-175 [CIP 104221]
Product TypeQuality Control Kit
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach