Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Choleraesuis ATCC™ 10708™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Choleraesuis ATCC™ 10708™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Choleraesuis ATCC™ 10708™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Choleraesuis ATCC™ 10708™
Thermo Scientific™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Choleraesuis ATCC™ 10708™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4719001100 Tests/Kit
Catalog number R4719001
Price (TWD)
-
Request A Quote
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Choleraesuis ATCC™ 10708™ is supplied as a film of preserved microorganisms on inside of cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®10708TM*
DescriptionSalmonella enterica subsp. enterica serovar Choleraesuis ATCC™ 10708™
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsETS 34 [CIP 104220]
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach