This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Convenience Set, Nutritional Adequacy Testing QC Set
Thermo Scientific™
Quanti-Cult Plus™ Convenience Set, Nutritional Adequacy Testing QC Set
Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Have Questions?
產品號碼Quantity
R4723000120 Tests/Set
產品號碼 R4723000
價格 (HKD)
-
Quantity:
120 Tests/Set
Thermo Scientific™ Quanti-Cult Plus Nutritional Adequacy Testing Set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
Includes:
12 preserved organism vials (two per organism); 12 rehydration vials
Contents:
Aspergillus brasiliensis ATCC™ 16404™
Bacillus spizizenii ATCC™ 6633 
Candida albicans ATCC™ 10231™
Escherichia coli ATCC™8739™
Pseudomonas paraeruginosa ATCC™ 9027™
Staphylococcus aureus subsp. aureus ATCC™ 6538™.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
規格
ATCC®16404™
DescriptionNutritional Adequacy Testing QC Set/6
FormatQuanti-Cult Plus
Packaging Type120 Test/Set
Quantity120 Tests/Set
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQuantitative QC
Unit SizeEach