Remel™ Reagent Rack
Product Image
Thermo Scientific™

Remel™ Reagent Rack

Thermo Scientific™ Remel Reagent Rack holds 2 x 50mL bottles and 7 x 25mL bottles.
Have Questions?
Catalog NumberHolds
R521901Bottles (2 x 50 mL and 7 x 25 mL)
Catalog number R521901
Price (USD)
-
Request A Quote
Holds:
Bottles (2 x 50 mL and 7 x 25 mL)

Thermo Scientific™ Remel Reagent Rack holds 2 x 50mL bottles and 7 x 25mL bottles.

4.5 x 10.25 x 1.5in.

Specifications
HoldsBottles (2 x 50 mL and 7 x 25 mL)
Product TypeReagent Rack
Unit SizeEach