Thermo Scientific™

Remel™ Calgiswab™ Type 3 Swab: Swab with Plastic-shaft

Catalog number: R6015003
Thermo Scientific™

Remel™ Calgiswab™ Type 3 Swab: Swab with Plastic-shaft

Catalog number: R6015003

Plastic-shaft swab for use with Thermo Scientific™ Remel Calgiswab Swab system. System designed for nasopharyngeal specimen collection. 

 
Catalog Number
R6015003
Unit Size
Each
Description
Plastic-shaft; 1/Envelope; 100/Pk.
Price (USD)
Full specifications
For Use With (Application)Healthcare
Format1/Envelop
MaterialPlastic (Shaft)
SwabConfigurationRemel Calgiswab
DescriptionPlastic-shaft; 1/Envelope; 100/Pk.
Packaging1/Envelope
ShaftPlastic
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
R6015003Full specifications
EachPlastic-shaft; 1/Envelope; 100/Pk.Request A Quote
For Use With (Application)Healthcare
Format1/Envelop
MaterialPlastic (Shaft)
SwabConfigurationRemel Calgiswab
DescriptionPlastic-shaft; 1/Envelope; 100/Pk.
Packaging1/Envelope
ShaftPlastic
Unit SizeEach
Showing 1 of 1
Manufacturer: Puritan

Figures